missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AlphaProTech Critical Cover™ ComforTech™ Coveralls
Lightweight, durable microporous material delivers protection against particles, biologicals, and fluids in controlled environments. Available in 3 different styles.
Supplier: AlphaProTech CVJ40225
Description
Material rapidly dissipates vapor and heat on a microscopic level which helps to keep wearer cool and comfortable throughout the workday.
- Material has static dissipative properties
- Exhibits an ultra-low particle count and 99.9% filtration efficiency
- Fluid and particulate barrier
- Serged seams
- Latex-free
- White
Specifications
Cleanroom Coveralls | |
Microporous Composite Fabric | |
Single-use | |
Elastic | |
Serged | |
Elastic wrist and ankle | |
29.75 in. | |
75.56 cm | |
For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency | |
93.98 cm | |
Class 10-100,000 (ISO 4-8) |
2X-Large | |
White | |
Elastic | |
Zipper | |
Non-sterile | |
Yes | |
31 in. | |
78.74 cm | |
37 in. | |
ComforTech™ |