missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AlphaProTech Critical Cover™ ComforTech™ Coveralls
Lightweight, durable microporous material delivers protection against particles, biologicals, and fluids in controlled environments. Available in 3 different styles.
$250.71 - $392.59
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | Microporous Composite Fabric |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Size | Price | Quantity | ||||
19-161-902B
|
AlphaProTech
CVJ40222 |
Elastic wrist and ankle | Medium |
Case of 25 Each for $281.97
|
|
||||
19-165-678
|
AlphaProTech
CVJ48323 |
Elastic wrist, ankle and back, attached AquaTrak™ boots | Large |
Case of 25 Each for $313.04
|
|
||||
19-811-005
|
AlphaProTech
CVJ4C928 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | 5X-Large |
Case of 25 Each for $392.59
|
|
||||
19-130-4225
|
AlphaProTech
CVJ4C922 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | Medium |
Case of 25 Each for $310.87
|
|
||||
19-130-4226
|
AlphaProTech
CVJ4C923 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | Large |
Case of 25 Each for $314.96
|
|
||||
19-161-902D
|
AlphaProTech
CVJ40224 |
Elastic wrist and ankle | X-Large |
Case of 25 Each for $257.47
|
|
||||
19-165-677
|
AlphaProTech
CVJ48322 |
Elastic wrist, ankle and back, attached AquaTrak™ boots | Medium |
Case of 25 Each for $313.04
|
|
||||
19-165-680
|
AlphaProTech
CVJ48325 |
Elastic wrist, ankle and back, attached AquaTrak™ boots | 2X-Large |
Case of 25 Each for $338.15
|
|
||||
19-161-902C
|
AlphaProTech
CVJ40223 |
Elastic wrist and ankle | Large |
Case of 25 Each for $281.97
|
|
||||
19-161-902H
|
AlphaProTech
CVJ40228 |
Elastic wrist and ankle | 5X-Large |
Case of 25 Each for $342.14
|
|
||||
19-130-4229
|
AlphaProTech
CVJ4C926 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | 3X-Large |
Case of 25 Each for $319.69
|
|
||||
19-161-902A
|
AlphaProTech
CVJ40221 |
Elastic wrist and ankle | Small |
Case of 25 Each for $281.97
|
|
||||
19-161-902E
|
AlphaProTech
CVJ40225 |
Elastic wrist and ankle | 2X-Large |
Case of 25 Each for $250.71
|
|
||||
19-165-682
|
AlphaProTech
CVJ48327 |
Elastic wrist, ankle and back, attached AquaTrak™ boots | 4X-Large |
Case of 25 Each for $376.14
|
|
||||
19-130-4230
|
AlphaProTech
CVJ4C927 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | 4X-Large |
Case of 25 Each for $370.45
|
|
||||
19-161-902G
|
AlphaProTech
CVJ40227 |
Elastic wrist and ankle | 4X-Large |
Case of 25 Each for $325.92
|
|
||||
19-161-902F
|
AlphaProTech
CVJ40226 |
Elastic wrist and ankle | 3X-Large |
Case of 25 Each for $310.58
|
|
||||
19-165-683
|
AlphaProTech
CVJ48328 |
Elastic wrist, ankle and back, attached AquaTrak™ boots | 5X-Large |
Case of 25 Each for $339.56
|
|
||||
19-130-4227
|
AlphaProTech
CVJ4C924 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | X-Large |
Case of 25 Each for $263.46
|
|
||||
19-130-4228
|
AlphaProTech
CVJ4C925 |
Elastic hood, wrist with attached AquaTrak™ boots and ankle | 2X-Large |
Case of 25 Each for $329.46
|
|
||||
19-169-4034
|
AlphaProTech
CVJ40229 |
Elastic wrist and ankle | 6X-Large |
Case of 25 Each for $289.88
|
|
||||
Description
Material rapidly dissipates vapor and heat on a microscopic level which helps to keep wearer cool and comfortable throughout the workday.
- Material has static dissipative properties
- Exhibits an ultra-low particle count and 99.9% filtration efficiency
- Fluid and particulate barrier
- Serged seams
- Latex-free
- White
Specifications
Cleanroom Coveralls | |
White | |
Elastic | |
Serged | |
Yes | |
ComforTech™ |
Microporous Composite Fabric | |
Single-use | |
Zipper | |
Non-sterile | |
For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency | |
Class 10-100,000 (ISO 4-8) |