AlphaProTech Critical Cover™ Coverall Face Shields
Good peripheral vision while acting as a shield against dangerous splashes in the work environment
Supplier: AlphaProTech 2808
Description
- Lightweight and easily fits over glasses
- Double-sided anti-fog coating
- Low distorting and 100% optically clear
- Soft polyurethane foam headband fits snugly against the forehead
- Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Specifications
Anti-fog | |
Fits easily over glasses | |
Elastic Comfort Band | |
Half-face |
7 mil shield | |
Clear | |
100/Cs. |