Thermo Scientific™ Cryo Gloves - Mid-Arm Length
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
Supplier:  Thermo Scientific™ 189562
 View more versions of this product
Catalog No. 15-466-313
Explore available promotions
Description
Description
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
Specifications
Specifications
Mid-Arm Length
Specific
Handling Samples from Ultra Low Temperature and Cryogenic Storage
35.6 cm
Blue
Large
14 in.
Product Suggestions
Product Suggestions