Thermo Scientific™ CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Supplier:  Thermo Scientific™ 50384422
 View more versions of this product
Catalog No. 03-337-7S
Explore available promotions
Description
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
  • Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
  • Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
  • Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Specifications
Specifications
Silver
16 CryoBoxes/Rack
Horizontal
Cryogenic Box Storage Rack, Horizontal
5.625 x 8.875 x 23.25 in. (22.5 x 59.0 x 14.3 cm)
Each
Stainless Steel