Thermo Scientific™ CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
$1164.73 - $4050.97
Specifications
Color Silver
Material Stainless Steel
View More Specs
Products 9
Catalog Number Mfr. No. Design Holds Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Design Holds Dimensions (L x W x H) Price Quantity  
12-566-195
View Documents
Thermo Scientific™
367000
Vertical 12 CryoBoxes/Rack 5.625 x 26.5 x 5.531 in. (14.3 x 67 x 14 cm)
Each for $2,241.40
15-350-107E
View Documents
Thermo Scientific™
50360004
Vertical 4 CryoBoxes/Rack 5.625 x 5.5 x 8.75 in. (14.3. x 14 x 22.5 cm)
Each for $1,164.73
Case of 4 Each for $4,658.92
15-350-107F
View Documents
Thermo Scientific™
50360009
Vertical 9 CryoBoxes/Rack 5.625 x 5.5 x 19.75 in. (14.3 x 14 x 50.2 cm)
Each for $1,641.90
Case of 4 Each for $6,567.60
03-337-7R
View Documents
Thermo Scientific™
50384322
Horizontal 12 CryoBoxes/Rack 5.625 x 8.875 x 17.5 in. (22.5 x 44.4 x 14.3 cm)
Case of 2 Each for $3,797.57
03-337-7S
View Documents
Thermo Scientific™
50384422
Horizontal 16 CryoBoxes/Rack 5.625 x 8.875 x 23.25 in. (22.5 x 59.0 x 14.3 cm)
Case of 2 Each for $4,050.97
15-350-107C
View Documents
Thermo Scientific™
DS50350004
Vertical 4 CryoBoxes/Rack 3.375 x 3.25 x 8.75 in. (8.4 x 8.3 x 22.5 cm)
Each for $1,402.33
15-350-107D
View Documents
Thermo Scientific™
DS50350009
Vertical 9 CryoBoxes/Rack 3.375 x 3.25 x 19.75 in. (8.4 x 8.3 x 50.2 cm)
Case for $1,605.10
03-337-7L
View Documents
Thermo Scientific™
DS50370004
Vertical 4 CryoBoxes/Rack 5.625 x 5.5 x 16 in. (14.3 x 14 x 40.6 cm)
Each for $1,998.07
22-273032
View Documents
Thermo Scientific™
DS50370007
Vertical 7 CryoBoxes/Rack 5.625 x 5.5 x 27.75 in. (14.3 x 14 x 70.5 cm)
Each for $2,230.47
Description
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
  • Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
  • Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
  • Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Specifications
Specifications
Silver
Stainless Steel
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'