Thermo Scientific™ CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Supplier: Thermo Scientific™ 50360004
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
- Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
- Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
- Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Specifications
Silver | |
4 CryoBoxes/Rack | |
Vertical | |
Cryogenic Box Storage Rack, Vertical |
5.625 x 5.5 x 8.75 in. (14.3. x 14 x 22.5 cm) | |
4/cs. | |
Stainless Steel |