Thermo Scientific™ CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Supplier: Thermo Scientific™ DS50370007
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
- Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
- Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
- Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Specifications
Silver | |
7 CryoBoxes/Rack | |
Stainless Steel |
5.625 x 5.5 x 27.75 in. (14.3 x 14 x 70.5 cm) | |
Vertical | |
Freezer Rack |