Thermo Scientific™ CryoPlus™ Vapor Phase Starter Packages: PROMO
CryoPlus™ Vapor Phase Starter Packages include the equipment needed for vapor phase, liquid nitrogen storage. All packages include a CryoPlus auto-fill LN2 system with racks, riser and hot gas bypass. Choose from 120V or 230V packages.
$29343.44 - $59170.58
Specifications
Type Auto-Fill LN2 Cryo
Certifications/Compliance CE, CFDA, cULus
Access Security Cabinet and Microprocessor Key Lock
Plug Type NEMA 5-15
Voltage 115 V
View More Specs
Products 3
Catalog Number Mfr. No. Description Capacity (Metric) Includes Price Quantity  
Catalog Number Mfr. No. Description Capacity (Metric) Includes Price Quantity  
13-987-213PM
View Documents
Thermo Scientific™
7400RAKHGBR
CryoPlus 1 Vapor Phase Starter Kit 90 L CryoPlus 1 (model 7400), hot gas bypass option, 4 storage racks, 3.5 inch riser N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-987-214PM
View Documents
Thermo Scientific™
7402RAKHGBR
CryoPlus 2 Vapor Phase Starter Kit 200 L CryoPlus 2 (model 7402), hot gas bypass option, 10 storage racks, 3.5 inch riser N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-762-390PM
View Documents
Thermo Scientific™
7404RAKHGBR
CryoPlus 3 Vapor Phase Starter Kit 340 L CryoPlus 3 (model 7404), hot gas bypass option, 17 storage racks, 3.5 inch riser N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Four capacities: 90L CryoPlus 1, 200L CryoPlus 2, 340L CryoPlus 3 and 552L CryoPlus 4
  • 2mL vial capacities: 6,318 (CryoPlus 1), 13,500 (CryoPlus 2), 23,000 (CryoPlus 3), 39,000 (CryoPlus 4)
  • Top-mounted control panel allow easy access to the microprocessor controller for programming
  • Security features: front-mounted key lock, microprocessor key lock and rear-mounted recessed power switch to prevent accidental power shut-off
  • 24 tricolor LEDs continually display actual liquid nitrogen level and high-level/low-level setpoints
  • Counterbalanced lid with 100% clearance allows each entry into the storage area for retrieval of samples while minimizing exposure time
  • Temperature sleeve incorporated, providing colder temperatures at the top and a more efficient vapor phase operation
  • Vacuum insulated with stainless steel interior
  • LN2 transfer hose included 
Specifications
Specifications
Auto-Fill LN2 Cryo
Cabinet and Microprocessor Key Lock
115 V
60 Hz
CE, CFDA, cULus
NEMA 5-15
115 V 60 Hz
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'