Crystal Violet (Certified Biological Stain), Fisher Chemical™
For use in histology, cytology and bacteriology (staining)
Supplier:  Thermo Fisher Scientific FLC58125
 View more versions of this product
Catalog No. C581-25
Explore available promotions
Description
Description
Specifications
Specifications
Crystal Violet
Pass Test
C25H30ClN3
Gentian Violet, Basic Violet 3
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Certified Biological Stain
42555
25 g
548-62-9
Report %
MFCD00011750
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.98
Glass Bottle
Green
Solid
Chemical Identifiers
548-62-9
407.986
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
MFCD00011750
Gentian Violet, Basic Violet 3
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Videos
Safety and Handling
Safety and Handling
SDS Documents