missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Crystal Violet (Certified Biological Stain), Fisher Chemical™
For use in histology, cytology and bacteriology (staining)
$325.67 - $530.42
Chemical Identifiers
CAS | 548-62-9 |
---|---|
Molecular Formula | C25H30ClN3 |
Molecular Weight (g/mol) | 407.986 |
MDL Number | MFCD00011750 |
InChI Key | ZXJXZNDDNMQXFV-UHFFFAOYSA-M |
Synonym | Gentian Violet, Basic Violet 3 |
PubChem CID | 11057 |
ChEBI | CHEBI:41688 |
IUPAC Name | [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
SMILES | CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
C581-25
|
Thermo Fisher Scientific
FLC58125 |
25 g | Glass Bottle |
Each for $325.67
Case of 6 Each for $1,954.02
|
|
||||
C581-100
|
Thermo Fisher Scientific
FLC581100 |
100 g | Glass Bottle |
Each for $530.42
Case of 6 Each for $3,182.52
|
|
||||
Chemical Identifiers
548-62-9 | |
407.986 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
11057 | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
C25H30ClN3 | |
MFCD00011750 | |
Gentian Violet, Basic Violet 3 | |
CHEBI:41688 | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Specifications
548-62-9 | |
Report % | |
MFCD00011750 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.98 | |
Glass Bottle | |
Green | |
Solid |
Pass Test | |
C25H30ClN3 | |
Gentian Violet, Basic Violet 3 | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
Certified Biological Stain | |
42555 | |
25 g | |
Crystal Violet |