missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cupric Sulfate CS, USP/EP Blue Primary Solution, Ricca Chemical

1 mL = 62.4 mg CuSO4.5H2O

Supplier:  Ricca Chemical Company 249532

 View more versions of this product

Catalog No. 2495-32


Explore available promotions

Description

Description

Specifications

Specifications

Cupric Sulfate
Blue Primary Solution
CuH10O9S
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
24463
EP/USP
1 L
Blue
7758-99-8,7647-01-0,7732-18-5
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
1 mL = 62.4 mg CuSO4·5H2O
CHEBI:31440
Amber Glass Bottle
Liquid

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper(2+) pentahydrate sulfate
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
SDS Documents

Documents

Promotions

Promotions