missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cupric Sulfate CS, USP/EP Blue Primary Solution, Ricca Chemical
1 mL = 62.4 mg CuSO4.5H2O
$172.81 - $486.66
Chemical Identifiers
CAS | 7758-99-8 |
---|---|
Molecular Formula | CuH10O9S |
Molecular Weight (g/mol) | 249.68 |
MDL Number | MFCD00149681 |
InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
PubChem CID | 24463 |
ChEBI | CHEBI:31440 |
IUPAC Name | copper(2+) pentahydrate sulfate |
SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
2495-4
|
Ricca Chemical Company
24954 |
120 mL | Amber Glass Bottle |
Each for $172.81
|
|
||||
2495-16
|
Ricca Chemical Company
249516 |
500 mL | Amber Glass Bottle |
Each for $278.35
|
|
||||
2495-32
|
Ricca Chemical Company
249532 |
1 L | Amber Glass Bottle |
Each for $486.66
|
|
||||
Chemical Identifiers
7758-99-8 | |
249.68 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
24463 | |
copper(2+) pentahydrate sulfate |
CuH10O9S | |
MFCD00149681 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
CHEBI:31440 | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Specifications
Blue | |
CuH10O9S | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
249.68 | |
24463 | |
EP/USP | |
120 mL | |
Cupric Sulfate, Blue Primary Solution |
7758-99-8,7647-01-0,7732-18-5 | |
MFCD00149681 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
copper(2+) pentahydrate sulfate | |
1 mL = 62.4 mg CuSO4·5H2O | |
CHEBI:31440 | |
Amber Glass Bottle | |
Liquid |