missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cupric Sulfate CS, USP/EP Blue Primary Solution, Ricca Chemical
1 mL = 62.4 mg CuSO4.5H2O
Supplier: Ricca Chemical Company 24954
Specifications
Cupric Sulfate | |
Blue Primary Solution | |
CuH10O9S | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
249.68 | |
24463 | |
EP/USP | |
120 mL |
Blue | |
7758-99-8,7647-01-0,7732-18-5 | |
MFCD00149681 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
copper(2+) pentahydrate sulfate | |
1 mL = 62.4 mg CuSO4·5H2O | |
CHEBI:31440 | |
Amber Glass Bottle | |
Liquid |
Chemical Identifiers
7758-99-8 | |
249.68 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
24463 | |
copper(2+) pentahydrate sulfate |
CuH10O9S | |
MFCD00149681 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
CHEBI:31440 | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |