missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanamide, 50% w/w aq. soln., stab., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L204480B

 View more versions of this product

Catalog No. AAL204480B


Explore available promotions

Description

Description

It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

Solubility
Soluble in water (85 g/100 ml (25°C). Soluble in organic solvents.

Notes
May form precipitate on storage
Specifications

Specifications

Cyanamide
420-04-2
4.2
Odorless
1.395
MFCD00007572
UN2922
Light sensitive; May form a precipitate
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
XZMCDFZZKTWFGF-UHFFFAOYSA-N
cyanamide
9864
42.04
stabilized
1.06
None
CH2N2
H2NCN
1000 g
1732569
14,2684
Soluble in water (85g/100 ml (25°C). Soluble in organic solvents.
C(#N)N
42.041
CHEBI:16698
50% w/w aq. soln.

Chemical Identifiers

420-04-2
42.041
XZMCDFZZKTWFGF-UHFFFAOYSA-N
9864
cyanamide
CH2N2
MFCD00007572
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
CHEBI:16698
C(#N)N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H312-H319-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301+H311-H314-H317-H335-H351-H361fd-H373

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 206-992-3

RTECSNumber : GS5950000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only