Learn More
Cyanamide, 50% w/w aq. soln., stab., Thermo Scientific Chemicals
$62.18 - $284.23
Chemical Identifiers
CAS | 420-04-2 |
---|---|
Molecular Formula | CH2N2 |
Molecular Weight (g/mol) | 42.041 |
MDL Number | MFCD00007572 |
InChI Key | XZMCDFZZKTWFGF-UHFFFAOYSA-N |
Synonym | hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex |
PubChem CID | 9864 |
ChEBI | CHEBI:16698 |
IUPAC Name | cyanamide |
SMILES | C(#N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL2044822
|
Thermo Scientific Chemicals
L2044822 |
100 g |
Each for $62.18
|
|
||||||
AAL2044830
|
Thermo Scientific Chemicals
L2044830 |
250 g |
Each for $93.36
|
|
||||||
AAL204480B
|
Thermo Scientific Chemicals
L204480B |
1000 g |
Each for $284.23
|
|
||||||
Description
It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.
Solubility
Soluble in water (85 g/100 ml (25°C). Soluble in organic solvents.
Notes
May form precipitate on storage
Chemical Identifiers
420-04-2 | |
42.041 | |
XZMCDFZZKTWFGF-UHFFFAOYSA-N | |
9864 | |
cyanamide |
CH2N2 | |
MFCD00007572 | |
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex | |
CHEBI:16698 | |
C(#N)N |
Specifications
420-04-2 | |
4.2 | |
Odorless | |
1.395 | |
MFCD00007572 | |
UN2922 | |
Light sensitive; May form a precipitate | |
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex | |
XZMCDFZZKTWFGF-UHFFFAOYSA-N | |
cyanamide | |
9864 | |
42.04 | |
Cyanamide, stabilized |
1.06 | |
None | |
CH2N2 | |
H2NCN | |
100 g | |
1732569 | |
14,2684 | |
Soluble in water (85g/100 ml (25°C). Soluble in organic solvents. | |
C(#N)N | |
42.041 | |
CHEBI:16698 | |
50% w/w aq. soln. |
Safety and Handling
GHS H Statement
H301-H314-H312-H319-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301+H311-H314-H317-H335-H351-H361fd-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 206-992-3
RTECSNumber : GS5950000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only