Learn More
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110890010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Cyanuric chloride | |
108-77-0 | |
Beige-Brown to Yellow | |
190.0°C | |
Authentic | |
Glass bottle | |
MFCD00006046 | |
26,35 | |
1.92 | |
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine | |
MGNCLNQXLYJVJD-UHFFFAOYSA-N | |
2,4,6-trichloro-1,3,5-triazine | |
7954 | |
CHEBI:58964 | |
0.99 |
0.99 | |
145.0°C to 148.0°C | |
1.9200g/mL | |
>200°C | |
0.99 | |
C3Cl3N3 | |
1 kg | |
03,72; 04,522; 05,687; 06,149; 10,114; 15,105 | |
152691 | |
Solubility in water: reacts. Other solubilities: reacts with ethanol, 250g/L acetone (25°C), soluble in chloroform, hot ether, dioxane, ketones, and carbontetrachloride | |
ClC1=NC(Cl)=NC(Cl)=N1 | |
184.40 | |
0.57 mPa.s (150°C) | |
184.41 | |
Solidified Mass or Fragments |
Chemical Identifiers
108-77-0 | |
184.40 | |
MGNCLNQXLYJVJD-UHFFFAOYSA-N | |
7954 | |
ClC1=NC(Cl)=NC(Cl)=N1 |
C3Cl3N3 | |
MFCD00006046 | |
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine | |
CHEBI:58964 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
Harmful if swallowed.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 203-614-9
RTECSNumber : XZ1400000
TSCA : TSCA
RUO – Research Use Only