missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanuric chloride, 99%, Thermo Scientific Chemicals

$53.27 - $121.33

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
PubChem CID 7954
ChEBI CHEBI:58964
SMILES ClC1=NC(Cl)=NC(Cl)=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110892500
SDS View Documents
Thermo Scientific Chemicals
110892500
250 g Glass bottle
Each for $53.27
 
AC110890010
SDS View Documents
Thermo Scientific Chemicals
110890010
1 kg Glass bottle
Each for $121.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

108-77-0
184.40
MGNCLNQXLYJVJD-UHFFFAOYSA-N
7954
ClC1=NC(Cl)=NC(Cl)=N1
C3Cl3N3
MFCD00006046
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
CHEBI:58964

Specifications

108-77-0
Beige-Brown to Yellow
190.0°C
Authentic
Glass bottle
MFCD00006046
26,35
1.92
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
MGNCLNQXLYJVJD-UHFFFAOYSA-N
trichloro-1,3,5-triazine
7954
CHEBI:58964
0.99
Cyanuric chloride, 0.99
145.0°C to 148.0°C
1.9200g/mL
>200°C
0.99
C3Cl3N3
250 g
03,72; 04,522; 05,687; 06,149; 10,114; 15,105
152691
Solubility in water: reacts. Other solubilities: reacts with ethanol, 250g/L acetone (25°C), soluble in chloroform, hot ether, dioxane, ketones, and carbontetrachloride
ClC1=NC(Cl)=NC(Cl)=N1
184.40
0.57 mPa.s (150°C)
184.41
Solidified Mass or Fragments
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
Harmful if swallowed.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-614-9

RTECSNumber : XZ1400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only