missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclobutylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 209110010

 View more versions of this product

Catalog No. AC209110010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclobutylamine
Colorless to Yellow
81.0°C to 82.0°C
Authentic
Glass bottle
1.4366 to 1.4386
MFCD00001328
12, 4
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
C1CC(C1)N
71.11
71.11
Liquid
2516-34-9
0.8300g/mL
−4°C
97.5% min. (GC)
C4H9N
C4H7NH2
1 mL
0.83
KZZKOVLJUKWSKX-UHFFFAOYSA-N
cyclobutanamine
75645
98%

Chemical Identifiers

2516-34-9
71.11
KZZKOVLJUKWSKX-UHFFFAOYSA-N
75645
C1CC(C1)N
C4H9N
MFCD00001328
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
cyclobutanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Danger

EINECSNumber : 219-736-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only