Learn More
Cyclobutylamine, 98%, Thermo Scientific Chemicals
$97.45 - $331.06
Chemical Identifiers
CAS | 2516-34-9 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.11 |
MDL Number | MFCD00001328 |
InChI Key | KZZKOVLJUKWSKX-UHFFFAOYSA-N |
Synonym | cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride |
PubChem CID | 75645 |
IUPAC Name | cyclobutanamine |
SMILES | C1CC(C1)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC209110010
|
Thermo Scientific Chemicals
209110010 |
1 mL | Glass bottle |
Each for $97.45
|
|
||||
AC209110050
|
Thermo Scientific Chemicals
209110050 |
5 mL | Glass bottle |
Each for $331.06
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2516-34-9 | |
71.11 | |
KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
75645 | |
C1CC(C1)N |
C4H9N | |
MFCD00001328 | |
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
cyclobutanamine |
Specifications
2516-34-9 | |
0.8300g/mL | |
−4°C | |
97.5% min. (GC) | |
C4H9N | |
C4H7NH2 | |
1 mL | |
0.83 | |
KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
cyclobutanamine | |
75645 | |
98% | |
Cyclobutylamine |
Colorless to Yellow | |
81.0°C to 82.0°C | |
Authentic | |
Glass bottle | |
1.4366 to 1.4386 | |
MFCD00001328 | |
12, 4 | |
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
C1CC(C1)N | |
71.11 | |
71.11 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
EINECSNumber : 219-736-
RUO – Research Use Only