missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, 99+%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111110050

 View more versions of this product

Catalog No. AC111110050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexane
110-82-7
81.0°C
−18°C
Liquid
0.005% max.
0.77
C6H12
Vapor Pressure: 104mbar at 20°C
0.05% max. (K.F.)
15, 2716
Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
C1CCCCC1
84.16
CHEBI:29005
≥99%
6.5°C
0.7700g/mL
5 L
Plastic Drum
1.4250 to 1.4270
99+%
C6H12
MFCD00003814
0.94 mPa.s (20°C)
05, 20
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.15
Pure

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only