Learn More
Cyclohexane, 99+%, pure
CAS: 110-82-7 | C6H12 | 84.16 g/mol
$140.45 - $1137.42
Chemical Identifiers
| CAS | 110-82-7 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00003814 |
| InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
| Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
| PubChem CID | 8078 |
| ChEBI | CHEBI:29005 |
| IUPAC Name | cyclohexane |
| SMILES | C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC111110010
|
Thermo Scientific Chemicals
111110010 |
1 L | Glass Bottle |
Each for $140.45
Case of 4 Each for $0.00
|
|
||||
|
AC111110025
|
Thermo Scientific Chemicals
111110025 |
2.5 L | Glass Bottle |
Each for $267.55
Case of 4 Each for $0.00
|
|
||||
|
AC111110050
|
Thermo Scientific Chemicals
111110050 |
5 L | Plastic Drum |
Each for $539.90
|
|
||||
|
AC111110250
|
Thermo Scientific Chemicals
111110250 |
25 L | Plastic Drum |
Each for $1,137.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Specifications
| 6.5°C | |
| 0.7700g/mL | |
| 1 L | |
| Glass Bottle | |
| 1.4250 to 1.4270 | |
| 99+% | |
| C6H12 | |
| MFCD00003814 | |
| 0.94 mPa.s (20°C) | |
| 05, 20 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.15 | |
| Pure |
| 110-82-7 | |
| 81.0°C | |
| −18°C | |
| Liquid | |
| 0.005% max. | |
| 0.77 | |
| C6H12 | |
| Vapor Pressure: 104mbar at 20°C | |
| 0.05% max. (K.F.) | |
| 15, 2716 | |
| Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4. | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99% | |
| Cyclohexane |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/ph
Danger
RUO – Research Use Only