missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanone, 99.8%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111190250

 View more versions of this product

Catalog No. AC111190250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexanone
108-94-1
155.0°C
43°C
Glass Bottle
1.4450 to 1.4560
0.947
C6H10(=O)
0.2% max. (K.F.)
01,178
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
JHIVVAPYMSGYDF-UHFFFAOYSA-N
cyclohexanone
7967
98.14
Extra Pure
-47.0°C
0.9470g/mL
25 mL
Authentic
Liquid
99.8%
C6H10O
MFCD00001625
07, 8
15, 2719
Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents
C1CCC(=O)CC1
98.145
CHEBI:17854
99.8%

Chemical Identifiers

108-94-1
98.145
JHIVVAPYMSGYDF-UHFFFAOYSA-N
7967
cyclohexanone
C6H10O
MFCD00001625
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
CHEBI:17854
C1CCC(=O)CC1
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only