Learn More
Cyclohexanone, 99.8%, extra pure, Thermo Scientific Chemicals
$70.22 - $208.44
Chemical Identifiers
CAS | 108-94-1 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.145 |
MDL Number | MFCD00001625 |
InChI Key | JHIVVAPYMSGYDF-UHFFFAOYSA-N |
Synonym | ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon |
PubChem CID | 7967 |
ChEBI | CHEBI:17854 |
IUPAC Name | cyclohexanone |
SMILES | C1CCC(=O)CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC111190250
|
Thermo Scientific Chemicals
111190250 |
25 mL | Glass Bottle |
Each for $70.22
|
|
||||
AC111190010
|
Thermo Scientific Chemicals
111190010 |
1 L | Glass Bottle |
Each for $93.68
Case of 4 Each for $0.00
|
|
||||
AC111190025
|
Thermo Scientific Chemicals
111190025 |
2.5 L | Glass Bottle |
Each for $208.44
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-94-1 | |
98.145 | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
7967 | |
cyclohexanone |
C6H10O | |
MFCD00001625 | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
CHEBI:17854 | |
C1CCC(=O)CC1 |
Specifications
-47.0°C | |
0.9470g/mL | |
25 mL | |
Authentic | |
Liquid | |
99.8% | |
C6H10O | |
MFCD00001625 | |
07, 8 | |
15, 2719 | |
Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents | |
C1CCC(=O)CC1 | |
98.145 | |
CHEBI:17854 | |
99.8% | |
Cyclohexanone |
108-94-1 | |
155.0°C | |
43°C | |
Glass Bottle | |
1.4450 to 1.4560 | |
0.947 | |
C6H10(=O) | |
0.2% max. (K.F.) | |
01,178 | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
cyclohexanone | |
7967 | |
98.14 | |
Extra Pure |
Safety and Handling
GHS P Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only