missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone oxime, 97%

CAS: 100-64-1 | C6H11NO | 113.16 g/mol

$43.45 - $43.45

Chemical Identifiers

CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111201000
SDS View Documents
Thermo Scientific Chemicals
111201000
100 g Plastic Bottle
Each for $43.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-64-1
113.16
VEZUQRBDRNJBJY-UHFFFAOYSA-N
7517
ON=C1CCCCC1
C6H11NO
MFCD00001660
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
N-cyclohexylidenehydroxylamine

Specifications

88°C to 92°C
Beige-Brown to White
Authentic
Crystalline Powder, Crystals, or Chunks
C6H11NO
MFCD00001660
10, 2720
VEZUQRBDRNJBJY-UHFFFAOYSA-N
N-cyclohexylidenehydroxylamine
7517
≥96.0%
100-64-1
100 g
Plastic Bottle
97%
C6H10(=NOH)
07, 10
cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
ON=C1CCCCC1
113.16
113.16
Cyclohexanone oxime, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-874-0

RTECSNumber : GW1925000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only