Learn More
Cyclohexylamine hydrochloride, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 4998-76-9 |
---|---|
Molecular Formula | C6H14ClN |
Molecular Weight (g/mol) | 135.64 |
MDL Number | MFCD00050540 |
InChI Key | ZJUGSKJHHWASAF-UHFFFAOYSA-N |
Synonym | cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 |
PubChem CID | 2723770 |
IUPAC Name | cyclohexanamine;hydrochloride |
SMILES | [Cl-].[NH3+]C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC134655000
|
Thermo Scientific Chemicals
134655000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
4998-76-9 | |
135.64 | |
ZJUGSKJHHWASAF-UHFFFAOYSA-N | |
2723770 | |
[Cl-].[NH3+]C1CCCCC1 |
C6H14ClN | |
MFCD00050540 | |
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 | |
cyclohexanamine;hydrochloride |
Specifications
4998-76-9 | |
Authentic | |
Plastic bottle | |
MFCD00050540 | |
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 | |
ZJUGSKJHHWASAF-UHFFFAOYSA-N | |
cyclohexanamine;hydrochloride | |
2723770 | |
99% |
306.0°C | |
99% | |
C6H14ClN | |
500g | |
Solubility in water: 83 g/100mL water (17°C). Other solubilities: soluble in most common organic solvents, practically insoluble in ethers | |
[Cl-].[NH3+]C1CCCCC1 | |
135.64 | |
135.63 | |
Cyclohexylamine hydrochloride, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Suspected of damaging fertility.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 225-661-4
RTECSNumber : GX1440000
TSCA : TSCA
RUO â Research Use Only