missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexylbenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111291000

 View more versions of this product

Catalog No. AC111291000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexylbenzene
827-52-1
Colorless
238°C to 240°C
Authentic
Glass bottle
1.5250 to 1.527
MFCD00001451
05, 503
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
IGARGHRYKHJQSM-UHFFFAOYSA-N
cyclohexylbenzene
13229
98%
98%
5°C
0.9500g/mL
98°C
97.5% min. (GC)
C12H16
C6H5C6H11
100 mL
0.95
Solubility in water: insoluble
C1CCC(CC1)C1=CC=CC=C1
160.26
160.26
Liquid

Chemical Identifiers

827-52-1
160.26
IGARGHRYKHJQSM-UHFFFAOYSA-N
13229
C1CCC(CC1)C1=CC=CC=C1
C12H16
MFCD00001451
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
cyclohexylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-572-0

RTECSNumber : CZ1330000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only