Learn More
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
$124.92 - $337.04
Chemical Identifiers
CAS | 827-52-1 |
---|---|
Molecular Formula | C12H16 |
Molecular Weight (g/mol) | 160.26 |
MDL Number | MFCD00001451 |
InChI Key | IGARGHRYKHJQSM-UHFFFAOYSA-N |
Synonym | phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar |
PubChem CID | 13229 |
IUPAC Name | cyclohexylbenzene |
SMILES | C1CCC(CC1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC111291000
|
Thermo Scientific Chemicals
111291000 |
100 mL | Glass bottle |
Each for $124.92
|
|
||||
AC111295000
|
Thermo Scientific Chemicals
111295000 |
500 mL | Glass bottle |
Each for $337.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
827-52-1 | |
160.26 | |
IGARGHRYKHJQSM-UHFFFAOYSA-N | |
13229 | |
C1CCC(CC1)C1=CC=CC=C1 |
C12H16 | |
MFCD00001451 | |
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar | |
cyclohexylbenzene |
Specifications
827-52-1 | |
Colorless | |
238°C to 240°C | |
Authentic | |
Glass bottle | |
1.5250 to 1.527 | |
MFCD00001451 | |
05, 503 | |
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar | |
IGARGHRYKHJQSM-UHFFFAOYSA-N | |
cyclohexylbenzene | |
13229 | |
98% | |
Cyclohexylbenzene, 98% |
5°C | |
0.9500g/mL | |
98°C | |
97.5% min. (GC) | |
C12H16 | |
C6H5C6H11 | |
100 mL | |
0.95 | |
Solubility in water: insoluble | |
C1CCC(CC1)C1=CC=CC=C1 | |
160.26 | |
160.26 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-572-0
RTECSNumber : CZ1330000
TSCA : TSCA
RUO – Research Use Only