missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclooctanemethanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2045006

 View more versions of this product

Catalog No. AAB2045006


Description

Description

Cyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
Specifications

Specifications

Cyclooctanemethanol
0.94
C9H18O
5 g
It is soluble in water.
C1CCCC(CCC1)CO
142.242
142.24
3637-63-6
93°C (199°F)
MFCD00001749
cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane
ZHPBLHYKDKSZCQ-UHFFFAOYSA-N
cyclooctylmethanol
77196
97%

Chemical Identifiers

3637-63-6
142.242
ZHPBLHYKDKSZCQ-UHFFFAOYSA-N
77196
C1CCCC(CCC1)CO
C9H18O
MFCD00001749
cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane
cyclooctylmethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 222-862-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only