Learn More
Cyclooctanemethanol, 97%, Thermo Scientific Chemicals
$242.03 - $242.03
Chemical Identifiers
CAS | 3637-63-6 |
---|---|
Molecular Formula | C9H18O |
Molecular Weight (g/mol) | 142.242 |
MDL Number | MFCD00001749 |
InChI Key | ZHPBLHYKDKSZCQ-UHFFFAOYSA-N |
Synonym | cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane |
PubChem CID | 77196 |
IUPAC Name | cyclooctylmethanol |
SMILES | C1CCCC(CCC1)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2045006
|
Thermo Scientific Chemicals
B2045006 |
5 g |
Each for $242.03
|
|
|||||
Description
Cyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
Chemical Identifiers
3637-63-6 | |
142.242 | |
ZHPBLHYKDKSZCQ-UHFFFAOYSA-N | |
77196 | |
C1CCCC(CCC1)CO |
C9H18O | |
MFCD00001749 | |
cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane | |
cyclooctylmethanol |
Specifications
3637-63-6 | |
93°C (199°F) | |
MFCD00001749 | |
cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane | |
ZHPBLHYKDKSZCQ-UHFFFAOYSA-N | |
cyclooctylmethanol | |
77196 | |
97% |
0.94 | |
C9H18O | |
5 g | |
It is soluble in water. | |
C1CCCC(CCC1)CO | |
142.242 | |
142.24 | |
Cyclooctanemethanol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 222-862-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only