missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene oxide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 348520250

 View more versions of this product

Catalog No. AC348520250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclopentene oxide
0.9640g/mL
10°C
97.5% min. (GC)
C5H8O
MFCD00005161
17,21
cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
C1CC2OC2C1
84.12
84.12
285-67-6
102.0°C
Authentic
Glass bottle
1.4330 to 1.4350
25g
0.964
GJEZBVHHZQAEDB-UHFFFAOYNA-N
6-oxabicyclo[3.1.0]hexane
9244
98%

Chemical Identifiers

285-67-6
84.12
GJEZBVHHZQAEDB-UHFFFAOYNA-N
9244
C1CC2OC2C1
C5H8O
MFCD00005161
cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
6-oxabicyclo[3.1.0]hexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 206-005-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only