missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentyl bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111602500

 View more versions of this product

Catalog No. AC111602500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopentyl bromide
137-43-9
1.3900g/mL
35°C
97.5% min. (GC)
C5H9Br
C5H9Br
250 mL
1.39
Solubility in water: insoluble in water
C1CCC(C1)Br
149.03
149.03
Liquid
98%
Yellow to Brown
137°C to 139°C
Authentic
Glass bottle
1.4881 to 1.4901
MFCD00001359
05, 19
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
BRTFVKHPEHKBQF-UHFFFAOYSA-N
bromocyclopentane
8728
98%

Chemical Identifiers

137-43-9
149.03
BRTFVKHPEHKBQF-UHFFFAOYSA-N
8728
C1CCC(C1)Br
C5H9Br
MFCD00001359
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
bromocyclopentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-294-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only