missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl bromide, 98%

CAS: 137-43-9 | C5H9Br | 149.03 g/mol

$83.71 - $274.57

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111600500
SDS View Documents
Thermo Scientific Chemicals
111600500
50 mL Glass bottle
Each for $83.71
Only null left
 
AC111602500
SDS View Documents
Thermo Scientific Chemicals
111602500
250 mL Glass bottle
Each for $274.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

137-43-9
149.03
BRTFVKHPEHKBQF-UHFFFAOYSA-N
8728
C1CCC(C1)Br
C5H9Br
MFCD00001359
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
bromocyclopentane

Specifications

137-43-9
1.3900g/mL
35°C
97.5% min. (GC)
C5H9Br
C5H9Br
50 mL
1.39
Solubility in water: insoluble in water
C1CCC(C1)Br
149.03
149.03
Liquid
Yellow to Brown
137°C to 139°C
Authentic
Glass bottle
1.4881 to 1.4901
MFCD00001359
05, 19
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
BRTFVKHPEHKBQF-UHFFFAOYSA-N
bromocyclopentane
8728
98%
Cyclopentyl bromide, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-294-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only