missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentyl iodide, 98%, stabilized over copper, Thermo Scientific™

Chemical Identifiers

CAS 1556-18-9
Molecular Formula C5H9I
Molecular Weight (g/mol) 196.03
MDL Number MFCD00039446
InChI Key PCEBAZIVZVIQEO-UHFFFAOYSA-N
Synonym cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
PubChem CID 73784
IUPAC Name iodocyclopentane
SMILES IC1CCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC381730250
SDS View Documents
Thermo Scientific Chemicals
381730250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1556-18-9
196.03
PCEBAZIVZVIQEO-UHFFFAOYSA-N
73784
IC1CCCC1
C5H9I
MFCD00039446
cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
iodocyclopentane

Specifications

1556-18-9
166.0°C to 167.0°C
Authentic
Glass bottle
C5H9I
25mL
cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
IC1CCCC1
196.03
73784
98%
1.6950g/mL
52°C
96% min. (GC)
C5H9I
MFCD00039446
1.695
PCEBAZIVZVIQEO-UHFFFAOYSA-N
iodocyclopentane
(Copper)
196.03
Cyclopentyl iodide, Stabilized
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a

GHS Signal Word: Warning

EINECSNumber : 216-311-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only