missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Cyclopentyl isocyanide, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 372090010

 Afficher plus de versions de ce produit

Numéro de catalogue. AC372090010


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Cyclopentyl isocyanide
Colorless to Yellow
40°C (11.0 hPa)
95% min. (GC)
C6H9N
1 g
cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
XTLSDEJNOQVCHL-UHFFFAOYSA-N
isocyanocyclopentane
2734784
96%
68498-54-4
0.8600g/mL
Authentic
Glass bottle
1.4390 to 1.4410
0.86
Solubility in water: insoluble
[C-]#[N+]C1CCCC1
95.14
95.14
Liquid

Identifiants chimiques

68498-54-4
95.14
cyclopentyl isocyanide, cyclopentylisocyanide, isocyano-cyclopentan, isocyano-cyclopentane, cyclopentyl isonitrile, 3-isocyano-cyclopentane, acmc-1bgsw, tos-bb-0762
isocyanocyclopentane
C6H9N
XTLSDEJNOQVCHL-UHFFFAOYSA-N
2734784
[C-]#[N+]C1CCCC1
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and

GHS Signal Word: Warning

FDS Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only