missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 111631000

 View more versions of this product

Catalog No. AC111631000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopropanecarboxylic acid chloride
Colorless to Yellow
119.0°C
Authentic
Glass bottle
1.4515 to 1.4535
100 g
1.15
Solubility in water: decomposes
C1CC1C(=O)Cl
104.54
104.54
Liquid
4023-34-1
1.1500g/mL
23°C
97.5% min. (GC)
C4H5ClO
C3H5COCl
09,4
cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
ZOOSILUVXHVRJE-UHFFFAOYSA-N
cyclopropanecarbonyl chloride
77637
98%

Chemical Identifiers

4023-34-1
104.54
cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
cyclopropanecarbonyl chloride
C4H5ClO
ZOOSILUVXHVRJE-UHFFFAOYSA-N
77637
C1CC1C(=O)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 223-684-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only