missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic acid chloride, 98%

CAS: 4023-34-1 | C4H5ClO | 104.54 g/mol

$135.96 - $267.81

Chemical Identifiers

CAS 4023-34-1
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.54
InChI Key ZOOSILUVXHVRJE-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
PubChem CID 77637
IUPAC Name cyclopropanecarbonyl chloride
SMILES C1CC1C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111630250
SDS View Documents
Thermo Scientific Chemicals
111630250
25 g Glass bottle
Each for $135.96
Only null left
 
AC111631000
SDS View Documents
Thermo Scientific Chemicals
111631000
100 g Glass bottle
Each for $267.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4023-34-1
104.54
cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
cyclopropanecarbonyl chloride
C4H5ClO
ZOOSILUVXHVRJE-UHFFFAOYSA-N
77637
C1CC1C(=O)Cl

Specifications

4023-34-1
1.1500g/mL
23°C
97.5% min. (GC)
C4H5ClO
C3H5COCl
09,4
cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
ZOOSILUVXHVRJE-UHFFFAOYSA-N
cyclopropanecarbonyl chloride
77637
98%
Cyclopropanecarboxylic acid chloride
Colorless to Yellow
119.0°C
Authentic
Glass bottle
1.4515 to 1.4535
25 g
1.15
Solubility in water: decomposes
C1CC1C(=O)Cl
104.54
104.54
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 223-684-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only