missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopropyl 4-methoxyphenyl ketone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2072514

 View more versions of this product

Catalog No. AAB2072514


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclopropyl 4-methoxyphenyl ketone
40°C to 44°C
>110°C (230°F)
MFCD00001296
2046539
YKZSVEVTRUSPOQ-UHFFFAOYSA-N
cyclopropyl-(4-methoxyphenyl)methanone
81580
98%
7152-03-6
158°C to 160°C (4.5 mmHg)
C11H12O2
25 g
cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl
COC1=CC=C(C=C1)C(=O)C2CC2
176.22
176.22

Chemical Identifiers

7152-03-6
176.22
YKZSVEVTRUSPOQ-UHFFFAOYSA-N
81580
COC1=CC=C(C=C1)C(=O)C2CC2
C11H12O2
MFCD00001296
cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl
cyclopropyl-(4-methoxyphenyl)methanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 230-487-7

RTECSNumber : PC4946500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only