missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopropyl 4-methoxyphenyl ketone, 98%, Thermo Scientific Chemicals
$50.21 - $155.93
Chemical Identifiers
CAS | 7152-03-6 |
---|---|
Molecular Formula | C11H12O2 |
Molecular Weight (g/mol) | 176.22 |
MDL Number | MFCD00001296 |
InChI Key | YKZSVEVTRUSPOQ-UHFFFAOYSA-N |
Synonym | cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl |
PubChem CID | 81580 |
IUPAC Name | cyclopropyl-(4-methoxyphenyl)methanone |
SMILES | COC1=CC=C(C=C1)C(=O)C2CC2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2072506
|
Thermo Scientific Chemicals
B2072506 |
5 g |
Each for $50.21
|
|
|||||
AAB2072514
|
Thermo Scientific Chemicals
B2072514 |
25 g |
Each for $155.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7152-03-6 | |
176.22 | |
YKZSVEVTRUSPOQ-UHFFFAOYSA-N | |
81580 | |
COC1=CC=C(C=C1)C(=O)C2CC2 |
C11H12O2 | |
MFCD00001296 | |
cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl | |
cyclopropyl-(4-methoxyphenyl)methanone |
Specifications
7152-03-6 | |
158°C to 160°C (4.5 mmHg) | |
C11H12O2 | |
5 g | |
cyclopropyl 4-methoxyphenyl methanone, cyclopropyl 4-methoxyphenyl ketone, methanone, cyclopropyl 4-methoxyphenyl, chembl82673, cyclopropyl-4-methoxyphenyl methanone, cyclopropyl-4-methoxy-phenyl-methanone, maybridge3_006117, 4-methoxybenzoylcyclopropane, cyclopropyl p-anisyl ketone, methanone,cyclopropyl 4-methoxyphenyl | |
COC1=CC=C(C=C1)C(=O)C2CC2 | |
176.22 | |
176.22 | |
Cyclopropyl 4-methoxyphenyl ketone |
40°C to 44°C | |
>110°C (230°F) | |
MFCD00001296 | |
2046539 | |
YKZSVEVTRUSPOQ-UHFFFAOYSA-N | |
cyclopropyl-(4-methoxyphenyl)methanone | |
81580 | |
98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 230-487-7
RTECSNumber : PC4946500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only