Learn More
Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H51158AE
Description
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGrignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Cyclopropylmagnesium bromide | |
C3H5BrMg | |
100 mL | |
Air and moisture sensitive | |
Reacts with water. | |
Br[Mg]C1CC1 | |
145.28 | |
145.28 |
23719-80-4 | |
MFCD00672094 | |
UN3399 | |
cyclopropylmagnesium bromide, grignard reagent, bromo cyclopropyl magnesium, bromo cyclopropyl magnesium, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, cyclopropyl magnesiumbromide, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, cyclopropylmagnesiunn bromide | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
bromo(cyclopropyl)magnesium | |
11116189 | |
0.5M in MeTHF |
Chemical Identifiers
23719-80-4 | |
145.28 | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
11116189 |
C3H5BrMg | |
MFCD00672094 | |
cyclopropylmagnesium bromide, grignard reagent, bromo cyclopropyl magnesium, bromo cyclopropyl magnesium, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, cyclopropyl magnesiumbromide, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, cyclopropylmagnesiunn bromide | |
Br[Mg]C1CC1 |
Safety and Handling
GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335
DOTInformation : Hazard Class: 4.3; Packaging Group: I
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only