Learn More
Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
$370.05 - $579.14
Chemical Identifiers
CAS | 23719-80-4 |
---|---|
Molecular Formula | C3H5BrMg |
Molecular Weight (g/mol) | 145.28 |
MDL Number | MFCD00672094 |
InChI Key | QUPLLJGAJVXXBW-UHFFFAOYSA-M |
Synonym | cyclopropylmagnesium bromide, grignard reagent, bromo cyclopropyl magnesium, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cyclopropyl magnesiumbromide, cyclopropyl magnesiumbromide, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide |
PubChem CID | 11116189 |
SMILES | Br[Mg]C1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH51158AD
|
Thermo Scientific Chemicals
H51158AD |
50 mL |
N/A
|
|
|||||
AAH51158AE
|
Thermo Scientific Chemicals
H51158AE |
100 mL |
N/A
|
|
|||||
Description
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGrignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
23719-80-4 | |
145.28 | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
11116189 |
C3H5BrMg | |
MFCD00672094 | |
cyclopropylmagnesium bromide, grignard reagent, bromo cyclopropyl magnesium, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cyclopropyl magnesiumbromide, cyclopropyl magnesiumbromide, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide | |
Br[Mg]C1CC1 |
Specifications
23719-80-4 | |
MFCD00672094 | |
UN3399 | |
cyclopropylmagnesium bromide, grignard reagent, bromo cyclopropyl magnesium, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cyclopropyl magnesiumbromide, cyclopropyl magnesiumbromide, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide | |
QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
magnesium;cyclopropane;bromide | |
11116189 | |
0.5M in MeTHF |
C3H5BrMg | |
50 mL | |
Air and moisture sensitive | |
Reacts with water. | |
Br[Mg]C1CC1 | |
145.28 | |
145.28 | |
Cyclopropylmagnesium bromide |
Safety and Handling
GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335
DOTInformation : Hazard Class: 4.3; Packaging Group: I
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only