Synthware™ Cylindrical Stir Bars
PTFE magnetic stir bars feature a pivot ring, and are only intended for use with flat bottom flasks.
Supplier:  Synthware™ SB1255
 View more versions of this product
Catalog No. 31502347
Explore available promotions
Description
Description
  • PTFE magnetic stir bars
  • Cylindrical shape with pivot ring
  • For use with flat bottom flasks only
Specifications
Specifications
12 mm
Stirring Liquids
White
PTFE
Autoclavable
Cylinder
Magnetic
Stir Bar
1 Ea.
Flat Bottom Flasks
55 mm