Learn More
Cyproterone acetate, 98%, Thermo Scientific Chemicals
$157.45 - $157.45
Chemical Identifiers
CAS | 427-51-0 |
---|---|
Molecular Formula | C24H29ClO4 |
Molecular Weight (g/mol) | 416.94 |
InChI Key | UWFYSQMTEOIJJG-FDTZYFLXSA-N |
Synonym | cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 |
PubChem CID | 9880 |
ChEBI | CHEBI:50743 |
SMILES | CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC459542500
|
Thermo Scientific Chemicals
459542500 |
250 mg |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
427-51-0 | |
416.94 | |
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 | |
CHEBI:50743 |
C24H29ClO4 | |
UWFYSQMTEOIJJG-FDTZYFLXSA-N | |
9880 | |
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C |
Specifications
427-51-0 | |
0.1% max. | |
Authentic | |
C24H29ClO4 | |
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 | |
UWFYSQMTEOIJJG-FDTZYFLXSA-N | |
416.94 | |
CHEBI:50743 | |
98% |
207.0°C to 215.0°C | |
0.5% max. (80°C) | |
97.5% min. (HPLC) | |
250 mg | |
Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, methanol, slightly soluble in ethanol | |
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C | |
9880 | |
416.94 | |
Cyproterone acetate |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Warning
EINECSNumber : 207-048-3
RUO – Research Use Only