Learn More
Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For Research Use Only. Not for use in diagnostic procedures.
Supplier: Agilent BioTek CYT1VSN
Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Specifications
Cell Imaging Multi-Mode Microplate Reader | |
0 to 20% | |
Fluorescence and high contrast brightfield imaging | |
6-well to 384-well plates | |
Xenon Flash | |
Gen5 Microplate Reader and Imager Software | |
20 in. | |
17.5 in. | |
16.5 in. | |
CE and TUV marked. RoHS compliant. |
Linear, Orbital, Double Orbital | |
0 to 19% | |
CYT1V | |
Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process | |
0.3 μm/pixel at 20X | |
Incubation to 45°C | |
50.8 cm | |
44.5 cm | |
41.9 cm |