Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For Research Use Only. Not for use in diagnostic procedures.
$61524.69 - $86671.22
Specifications
Type Cell Imaging Multi-Mode Microplate Reader
Shaking Linear, Orbital, Double Orbital
CO2 Concentration Range 0 to 20%
O2 Concentration Range 0 to 19%
Plate Type 6-well to 384-well plates
View More Specs
Products 3
Catalog Number Mfr. No. Model Includes Price Quantity  
Catalog Number Mfr. No. Model Includes Price Quantity  
BTCYT1V
View Documents
Agilent BioTek
CYT1VSN
CYT1V Fluorescence and high contrast brightfield imaging N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT1FAV
View Documents
Agilent BioTek
CYT1FAVSN
CYT1FAV Fluorescence and high contrast brightfield imaging; Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTCYT1FA
View Documents
Agilent BioTek
CYT1FASN
CYT1FA Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Specifications
Specifications
Cell Imaging Multi-Mode Microplate Reader
0 to 20%
6-well to 384-well plates
Xenon Flash
Gen5 Microplate Reader and Imager Software
20 in.
17.5 in.
16.5 in.
CE and TUV marked. RoHS compliant.
Linear, Orbital, Double Orbital
0 to 19%
Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process
0.3 μm/pixel at 20X
Incubation to 45°C
50.8 cm
44.5 cm
41.9 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'