missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytochrome C From Horse Heart MP Biomedicals

$1,360.14 - $2,626.48

Chemical Identifiers

CAS 9007-43-6
Molecular Formula C40H48FeN6O6S4
Molecular Weight (g/mol) 892.94
MDL Number MFCD00130890
InChI Key UMUOLVQYJUIZKD-RHANTFMNSA-L
Synonym cytochrome c, 3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+
PubChem CID 16057918
IUPAC Name 3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+)
SMILES CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN1014675
View Documents
MP Biomedicals Inc
021014675
500 mg N/A
 
ICN10146701
View Documents
MP Biomedicals Inc
0210146701
1 g N/A
 
Description

Description

Specifications

Chemical Identifiers

9007-43-6
892.94
UMUOLVQYJUIZKD-RHANTFMNSA-L
16057918
CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C
C40H48FeN6O6S4
MFCD00130890
cytochrome c, 3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+
3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+)

Specifications

9007-43-6
MFCD00130890
cytochrome c, 3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+
CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C
892.94
12000
C40H48FeN6O6S4
500 mg
UMUOLVQYJUIZKD-RHANTFMNSA-L
3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+)
16057918
Cytochrome C, From Horse Heart
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'