missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6379503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-Aspartic acid 4-tert-butyl ester | |
C8H15NO4 | |
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid | |
CC(C)(C)OC(=O)CC(N)C(O)=O | |
1 g | |
7018828 | |
98% |
64960-75-4 | |
MFCD00077101 | |
MXWMFBYWXMXRPD-UHFFFAOYNA-N | |
(2R)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
189.21 | |
189.21 | |
Solid |
Chemical Identifiers
64960-75-4 | |
189.21 | |
MXWMFBYWXMXRPD-UHFFFAOYNA-N | |
7018828 |
C8H15NO4 | |
MFCD00077101 | |
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid | |
CC(C)(C)OC(=O)CC(N)C(O)=O |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold