missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific™

Chemical Identifiers

CAS 64960-75-4
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00077101
InChI Key MXWMFBYWXMXRPD-UHFFFAOYNA-N
Synonym h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
PubChem CID 7018828
SMILES CC(C)(C)OC(=O)CC(N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6379503
View Documents
Thermo Scientific Chemicals
H6379503
1 g N/A
 
AAH6379506
View Documents
Thermo Scientific Chemicals
H6379506
5 g N/A
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

64960-75-4
189.21
MXWMFBYWXMXRPD-UHFFFAOYNA-N
7018828
C8H15NO4
MFCD00077101
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
CC(C)(C)OC(=O)CC(N)C(O)=O

Specifications

64960-75-4
MFCD00077101
MXWMFBYWXMXRPD-UHFFFAOYNA-N
(2R)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
189.21
189.21
Solid
C8H15NO4
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
CC(C)(C)OC(=O)CC(N)C(O)=O
1 g
7018828
98%
D-Aspartic acid 4-tert-butyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'