missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6379506

 View more versions of this product

Catalog No. AAH6379506

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

D-Aspartic acid 4-tert-butyl ester
C8H15NO4
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
CC(C)(C)OC(=O)CC(N)C(O)=O
5 g
7018828
98%
64960-75-4
MFCD00077101
MXWMFBYWXMXRPD-UHFFFAOYNA-N
2-amino-4-(tert-butoxy)-4-oxobutanoic acid
189.21
189.21
Solid

Chemical Identifiers

64960-75-4
189.21
MXWMFBYWXMXRPD-UHFFFAOYNA-N
7018828
C8H15NO4
MFCD00077101
h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
CC(C)(C)OC(=O)CC(N)C(O)=O
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions