missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6319703

 View more versions of this product

Catalog No. AAH6319703


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

114°C to 119°C
69630-50-8
MFCD00070384
PNLXWGDXZOYUKB-PGMHMLKASA-N
dimethyl (2R)-2-aminobutanedioate;hydrochloride
197.615
197.62
Solid
D-Aspartic acid dimethyl ester hydrochloride
C6H12ClNO4
d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl
COC(=O)CC(C(=O)OC)N.Cl
1 g
51051594
98%

Chemical Identifiers

69630-50-8
197.615
PNLXWGDXZOYUKB-PGMHMLKASA-N
51051594
COC(=O)CC(C(=O)OC)N.Cl
C6H12ClNO4
MFCD00070384
d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl
dimethyl (2R)-2-aminobutanedioate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only